Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediamine, BAKER ANALYZED™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

107-15-3
C2H8N2
60.1
PIICEJLVQHRZGT-UHFFFAOYSA-N
3301
CHEBI:30347

High quality chemicals for laboratory and specialized industrial use

Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.89kg
SMILES C(CN)N
IUPAC Name ethane-1,2-diamine
Formula Weight 60.1
Quantity 500mL
Flash Point 34.00°C
Chemical Name or Material Ethylenediamine
Grade BAKER ANALYZED™ Reagent