Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediaminetetraacetic acid, copper(II) disodium salt tetrahydrate, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
39208-15-6
C10H20CuN2Na2O12
469.798
MFCD00661154
CHPMNDHAIUIBSK-UHFFFAOYSA-J
copper ii disodium ethylenediaminetetraacetate tetrahydrate,ethylenediaminetetraacetic acidcopper ii disodium salt tetrahydrate,acmc-209j3w,edta-cu-15,ksc911m0f,copper 2+ disodium ion 4-edta tetrahydrate,copper ii disodium 2,2',2,2'-ethane-1,2-diylbis azanetriyl tetraacetate tetrahydrate
46245299
copper;disodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;tetrahydrate
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.O.O.O.[Na+].[Na+].[Cu+2]
39208-15-6
C10H20CuN2Na2O12
469.798
MFCD00661154
CHPMNDHAIUIBSK-UHFFFAOYSA-J
copper ii disodium ethylenediaminetetraacetate tetrahydrate,ethylenediaminetetraacetic acidcopper ii disodium salt tetrahydrate,acmc-209j3w,edta-cu-15,ksc911m0f,copper 2+ disodium ion 4-edta tetrahydrate,copper ii disodium 2,2',2,2'-ethane-1,2-diylbis azanetriyl tetraacetate tetrahydrate
46245299
copper;disodium;2-[2-[bis(carboxylatomethyl)amino]ethyl-(carboxylatomethyl)amino]acetate;tetrahydrate
C(CN(CC(=O)[O-])CC(=O)[O-])N(CC(=O)[O-])CC(=O)[O-].O.O.O.O.[Na+].[Na+].[Cu+2]
Chelation/Complexation reagent
pH 7 to 9 (1% soln.)
Color Blue
Appearance (1% aq. soln.) clear, blue
Infrared Spectrum Authentic
Quantity 25g
Assay Percent Range 98%
Formula Weight 469.8
Physical Form Fine Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material Ethylenediaminetetraacetic acid, copper(II) disodium salt tetrahydrate, 98%

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 254-356-9