Alfa Aesar™ Ethylenediaminetetraacetic acid disodium salt, 0.115N (0.0575M) Standardized solution

Manufacturer: Alfa Aesar™ 39782K7

Quantity 4L

 View more versions of this product

Catalog No. AA39782K7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 139-33-3
    Formula Weight 336.22
    MDL Number MFCD00070672
    Synonym EDTA disodium salt
    Chemical Name or Material Ethylenediaminetetraacetic acid disodium salt, Standardized Solution

    Specifications

    Formula Weight 336.22
    Synonym EDTA disodium salt
    Quantity 4L
    Physical Form Liquid
    Concentration 0.115N (0.0575M)
    MDL Number MFCD00070672
    TSCA TSCA
    CAS 139-33-3
    Odor Odorless
    Chemical Name or Material Ethylenediaminetetraacetic acid disodium salt, Standardized Solution

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit