Alfa Aesar™ Ethylenediaminetetraacetic acid, iron(III) monosodium salt

Manufacturer: Alfa Aesar™ 8899536

Quantity 500g

 View more versions of this product

Catalog No. AA8899536

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15708-41-5
    Molecular Formula FeNa(O2CCH2)2NCH2CH2N(CH2CO2)2
    Formula Weight 367.05
    MDL Number MFCD00078215
    Chemical Name or Material Ethylenediaminetetraacetic acid, iron (III) monosodium salt

    Specifications

    Formula Weight 367.05
    Physical Form Powder
    MDL Number MFCD00078215
    CAS 15708-41-5
    Odor Odorless
    TSCA TSCA
    Molecular Formula FeNa(O2CCH2)2NCH2CH2N(CH2CO2)2
    Quantity 500g
    Chemical Name or Material Ethylenediaminetetraacetic acid, iron (III) monosodium salt

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit