Alfa Aesar™ Ethylenediaminetetraacetic acid, magnesium, disodium salt

Manufacturer: Alfa Aesar™ 3054818

Quantity 50g

 View more versions of this product

Catalog No. AA3054818

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 14402-88-1
    Formula Weight 358.51
    MDL Number MFCD00078217
    Chemical Name or Material Ethylenediaminetetraacetic acid, magnesium, disodium salt

    Specifications

    Formula Weight 358.51
    TSCA TSCA
    CAS 14402-88-1
    Quantity 50g
    Physical Form Powder
    MDL Number MFCD00078217
    Odor Odorless
    Chemical Name or Material Ethylenediaminetetraacetic acid, magnesium, disodium salt

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit