Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediaminetetraacetic Acid Magnesium Disodium Salt, Spectrum™

Click to view available options
Quantity:
100 g
250 g
1 kg
25 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
14402-88-1
14402-88-1
Packaging Poly Pail
Insoluble Matter 5e-05
Quantity 1 kg
CAS Max % 1
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.