Alfa Aesar™ Ethylenediaminetetraacetic acid manganese disodium salt hydrate

Metal chelating compound

Manufacturer: Alfa Aesar™ 4051906

Quantity 5g

 View more versions of this product

Catalog No. AA4051906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 15375-84-5
    Formula Weight 389.13 (Anhydrous)
    MDL Number MFCD00661156
    Chemical Name or Material Ethylenediaminetetraacetic acid manganese disodium salt hydrate

    Specifications

    Formula Weight 389.13 (Anhydrous)
    CAS 15375-84-5
    TSCA TSCA
    Physical Form Powder
    MDL Number MFCD00661156
    Odor Odorless
    Quantity 5g
    Chemical Name or Material Ethylenediaminetetraacetic acid manganese disodium salt hydrate

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit