Alfa Aesar™ Ethylenediaminetetraacetic acid tetrakis(tetramethylammonium) salt, 30% w/v aq. soln.

Chelating agent, polymerization catalyst, detergent

Manufacturer: Alfa Aesar™ 41518AK

Quantity 250mL

 View more versions of this product

Catalog No. AA41518AK

  Check Availability
Add to Cart

    Chemical Identifiers

    Formula Weight 276.44
    MDL Number MFCD01632148
    Synonym EDTA tetrakis(tetramethylammonium) salt; Tetrakis(tetramethylammonium) ethylenediaminetetraacetate
    Chemical Name or Material Ethylenediaminetetraacetic acid tetrakis(tetramethylammonium) salt

    Specifications

    Formula Weight 276.44
    Physical Form Liquid
    MDL Number MFCD01632148
    Synonym EDTA tetrakis(tetramethylammonium) salt; Tetrakis(tetramethylammonium) ethylenediaminetetraacetate
    Quantity 250mL
    Concentration 30% w/v aq. soln.
    Chemical Name or Material Ethylenediaminetetraacetic acid tetrakis(tetramethylammonium) salt

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit