Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylenediaminetetraacetic Acid Tetrasodium Salt, Dihydrate, Reagent, 98%, Spectrum™ Chemical
SDP

Catalog Number 18602571
Click to view available options
Quantity:
125 g
500 g
3 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
10378-23-1
C10H20N2Na4O10
420.23
DGWOWDJDIBPYOR-UHFFFAOYSA-N
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid dihydrate tetrasodium
O.O.[Na].[Na].[Na].[Na].OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O
10378-23-1
C10H20N2Na4O10
420.23
DGWOWDJDIBPYOR-UHFFFAOYSA-N
2-({2-[bis(carboxymethyl)amino]ethyl}(carboxymethyl)amino)acetic acid dihydrate tetrasodium
O.O.[Na].[Na].[Na].[Na].OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O

E1047, 10378-23-1, C10H12N2O8Na4•2H2O

Spectrum™ Chemical Ethylenediaminetetraacetic Acid Tetrasodium Salt, Dihydrate, Reagent is a solid that has no color and is water soluble. It is mainly used to dissolve lime scale but has many other uses in fields such as medicine, industry, cosmetics and laboratories as a chelating agent and sequestrant. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.
CAS Max % 100%
Packaging Poly Pail
Quantity 3 kg
Percent Purity 98%
Grade Reagent
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit