Alfa Aesar™ Ethylenediaminetetraacetic acid trisodium salt hydrate, 95%

Manufacturer: Alfa Aesar™ H5616530

Quantity 250g

 View more versions of this product

Catalog No. AAH5616530

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 85715-60-2
    Molecular Formula C10H13N2Na3O8·xH2O
    Formula Weight 358.19
    MDL Number MFCD00149675
    Synonym EDTA trisodium salt hydrate
    Chemical Name or Material Ethylenediaminetetraacetic acid trisodium salt hydrate

    Specifications

    Formula Weight 358.19
    MDL Number MFCD00149675
    TSCA TSCA
    Molecular Formula C10H13N2Na3O8·xH2O
    CAS 85715-60-2
    Synonym EDTA trisodium salt hydrate
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point >300°C
    Quantity 250g
    Chemical Name or Material Ethylenediaminetetraacetic acid trisodium salt hydrate
    Assay 95%

    Description

    Applications
    This is used to eliminate enzyme inhibition by traces of heavy metals, and to inhibit enzymes that require divalent cations as cofactors.

    Solubility
    Soluble in sodium chloride. (1 M NaOH).

    Notes
    Incompatible with oxidizing agents. Store at room temperature.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit