Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
(Ethylenedinitrilo)tetraacetic Acid Disodium, Dihydrate, Reagent, ACS, 99-101%, Spectrum™ Chemical
Click to view available options
Quantity:
125 g
500 g
2.5 kg
6 x 125 g
6 x 500 g
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
Description
Spectrum™ Chemical (Ethylenedinitrilo)tetraacetic Acid Disodium, Dihydrate, Reagent, ACS , is one of several salts produced from Ethylenediaminetetraacetic acid (EDTA), itself an aminopolycarboxylic acid. Widely used in laboratory conditions, its uses vary from scavenging metal ions to acting as a chelating agent that binds cells in tissue cultures. As an ACS grade Reagent, Spectrum Chemical manufactured (ETHYLENEDINITRILO)TETRAACETIC ACID DISODIUM, DIHYDRATE is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness.
Specifications
CAS Max % | 100% |
Packaging | Fiber Drum |
Quantity | 45 kg |
Percent Purity | 99 to 101% |
Grade | ACS |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction