Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl nipecotate, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
5006-62-2
C8H15NO2
157.21
MFCD00005991
XIWBSOUNZWSFKU-UHFFFAOYSA-N
ethyl nipecotate, ethyl 3-piperidinecarboxylate, nipecotic acid ethyl ester, 3-piperidinecarboxylic acid ethyl ester, ethylpiperidine-3-carboxylate, 3-ethoxycarbonyl piperidine, 3-piperidinecarboxylic acid, ethyl ester, piperidine-3-carboxylic acid ethyl ester, ethylnipecotate, a-ethyl nipecotate
98969
ethyl piperidine-3-carboxylate
CCOC(=O)C1CCCNC1
5006-62-2
C8H15NO2
157.21
MFCD00005991
XIWBSOUNZWSFKU-UHFFFAOYSA-N
ethyl nipecotate, ethyl 3-piperidinecarboxylate, nipecotic acid ethyl ester, 3-piperidinecarboxylic acid ethyl ester, ethylpiperidine-3-carboxylate, 3-ethoxycarbonyl piperidine, 3-piperidinecarboxylic acid, ethyl ester, piperidine-3-carboxylic acid ethyl ester, ethylnipecotate, a-ethyl nipecotate
98969
ethyl piperidine-3-carboxylate
CCOC(=O)C1CCCNC1
Flash Point 90°C
Boiling Point 102°C to 104°C (7.0mmHg)
Specific Gravity 1.01
Density 1.0100g/mL
Refractive Index 1.4580 to 1.461
Quantity 100g
Packaging Glass bottle
Color Undesignated
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Solubility Information Solubility in water: miscible. Other solubilities: soluble in most organic solvents
Formula Weight 157.21
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Ethyl nipecotate
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 225-681-3