Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethyl p-toluenesulfonate, 98%, Acros Organics™

Click to view available options
Quantity:
50g
250g
1kg
Packaging:
Glass bottle
80-40-0
C9H12O3S
200.25
MFCD00009100
VRZVPALEJCLXPR-UHFFFAOYSA-N
ethyl p-toluenesulfonate,ethyl tosylate,ethyl p-tosylate,ethyl p-ts,benzenesulfonic acid, 4-methyl-, ethyl ester,ethyl p-methylbenzenesulfonate,ethyl-p-toluenesulfonate,p-toluenesulfonic acid, ethyl ester,ethyl pts,p-toluolsulfonsaeure aethyl ester
6638
ethyl 4-methylbenzenesulfonate
CCOS(=O)(=O)C1=CC=C(C=C1)C
80-40-0
C9H12O3S
200.25
MFCD00009100
VRZVPALEJCLXPR-UHFFFAOYSA-N
ethyl p-toluenesulfonate,ethyl tosylate,ethyl p-tosylate,ethyl p-ts,benzenesulfonic acid, 4-methyl-, ethyl ester,ethyl p-methylbenzenesulfonate,ethyl-p-toluenesulfonate,p-toluenesulfonic acid, ethyl ester,ethyl pts,p-toluolsulfonsaeure aethyl ester
6638
ethyl 4-methylbenzenesulfonate
CCOS(=O)(=O)C1=CC=C(C=C1)C
Melting Point 32.0°C to 34.0°C
Specific Gravity 1.174
Boiling Point 158.0°C to 162.0°C (10.0mmHg)
Refractive Index 1.5100 to 1.5120
Infrared Spectrum Authentic
Density 1.1740g/mL
Packaging Glass bottle
Quantity 250g
Color Colorless to Brown
Flash Point 157°C
Assay Percent Range 98%
Linear Formula CH3C6H4SO3C2H5
Beilstein 11, 99
Merck Index 15, 3907
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in many organic solvents
Formula Weight 200.25
Physical Form Liquid After Melting
Percent Purity 98%
Chemical Name or Material Ethyl p-toluenesulfonate
Show More Show Less

GHS H Statement:
Suspected of causing genetic defects.
Suspected of causing cancer.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning