Alfa Aesar™ Ethylferrocene, 98%

Manufacturer: Alfa Aesar™ 3925302

Quantity 0.5g

 View more versions of this product

Catalog No. AA3925302

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1273-89-8
    Formula Weight 214.09
    MDL Number MFCD00059071
    Chemical Name or Material Ethylferrocene

    Specifications

    Formula Weight 214.09
    Physical Form Liquid
    MDL Number MFCD00059071
    Density 1.256g/mL
    Refractive Index 1.6010
    CAS 1273-89-8
    Boiling Point 264°C
    Quantity 0.5g
    TSCA TSCA
    Chemical Name or Material Ethylferrocene
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit