Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylferrocene, 98%, Acros Organics

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
1273-89-8
C12H14Fe
1273-89-8
C12H14Fe
Quantity 5g
Packaging Glass bottle
Assay Percent Range 98%
Formula Weight 214.09
Chemical Name or Material Ethylferrocene