Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethylhydrocupreine hydrochloride, 97%

Click to view available options
Quantity:
100mg
500mg
3413-58-9
C21H29ClN2O2
376.925
MFCD00135594
QNRATNLHPGXHMA-XZHTYLCXSA-N
ethylhydrocupreine hydrochloride,optoquin hydrochloride,ethylhydrocupreine hcl,optochin hydrochloride,unii-s3n0jb4fc7,neumolisina,numoquin hydrochloride,s3n0jb4fc7,optoquinhydrochloride,optochin
16219340
(R)-(6-ethoxyquinolin-4-yl)-[(2S,4S,5R)-5-ethyl-1-azabicyclo[2.2.2]octan-2-yl]methanol;hydrochloride
CCC1CN2CCC1CC2C(C3=C4C=C(C=CC4=NC=C3)OCC)O.Cl
3413-58-9
C21H29ClN2O2
376.925
MFCD00135594
QNRATNLHPGXHMA-XZHTYLCXSA-N
ethylhydrocupreine hydrochloride,optoquin hydrochloride,ethylhydrocupreine hcl,optochin hydrochloride,unii-s3n0jb4fc7,neumolisina,numoquin hydrochloride,s3n0jb4fc7,optoquinhydrochloride,optochin
16219340
(R)-(6-ethoxyquinolin-4-yl)-[(2S,4S,5R)-5-ethyl-1-azabicyclo[2.2.2]octan-2-yl]methanol;hydrochloride
CCC1CN2CCC1CC2C(C3=C4C=C(C=CC4=NC=C3)OCC)O.Cl
Quantity 500mg
Melting Point ~240°C (decomposition)
Formula Weight 374.91
Percent Purity 97%
Chemical Name or Material Ethylhydrocupreine hydrochloride

RTECSNumber : MW6300000

Recommended Storage : 2° to 8°C