Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethylidene diacetate, 99%, Thermo Scientific™
Click to view available options
Quantity:
500g
Packaging:
Glass Bottle
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Color | Colorless to Yellow |
Boiling Point | 69.0°C (17.0 mmHg) |
Flash Point | 68°C |
Assay Percent Range | 98.5% min. (GC) |
Infrared Spectrum | Authentic |
Linear Formula | CH3CH(OCOCH3)2 |
Packaging | Glass Bottle |
Merck Index | 15,3866 |
Refractive Index | 1.3995 to 1.4015 |
Quantity | 500g |
Show More |
Safety and Handling
EINECSNumber : 208-800-3
RUO – Research Use Only