Alfa Aesar™ Ethylmagnesium bromide, 3M in ether

Grignard reagent

Manufacturer: Alfa Aesar™ 87291JU

Quantity 11 mole

 View more versions of this product

Catalog No. AA87291JU

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 925-90-6
    Molecular Formula C2H5BrMg
    Linear Formula C2H5MgBr
    Formula Weight 133.28
    MDL Number MFCD00000043
    UN Number UN3399
    Chemical Name or Material Ethylmagnesium bromide

    Specifications

    Formula Weight 133.28
    Health Hazard 3 GHS P Statement
    P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    CAS 925-90-6
    Health Hazard 2 GHS H Statement
    H224-H260-H314-H336
    Extremely flammable liquid and vapor.
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes severe skin burns and eye damage.
    May cause drowsiness or dizziness.
    Melting Point −116.3°C
    Concentration 3M in ether
    Quantity 11 mole
    Molecular Formula C2H5BrMg
    MDL Number MFCD00000043
    Sensitivity Air & Moisture Sensitive
    DOT Information Transport Hazard Class: 4.3; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
    TSCA TSCA
    Physical Form Liquid
    Linear Formula C2H5MgBr
    Density 1.020g/mL
    Boiling Point 34.6°C
    Odor Ether-like
    UN Number UN3399
    Health Hazard 1 Danger
    Color Pale yellow to brown
    Chemical Name or Material Ethylmagnesium bromide

    GHS H Statement
    H224-H260-H314-H336
    Extremely flammable liquid and vapor.
    In contact with water releases flammable gases which may ignite spontaneously.
    Causes severe skin burns and eye damage.
    May cause drowsiness or dizziness.

    GHS P Statement
    P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Handle under inert gas.
    Protect from moisture.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit