Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylmagnesium bromide, ca. 3.2M solution in 2-MeTHF; AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass bottle
925-90-6,96-47-9
C2H5BrMg
133.271
MFCD00000043
FRIJBUGBVQZNTB-UHFFFAOYSA-M
ethylmagnesium bromide,ethyl magnesium bromide,magnesium, bromoethyl,ethylmagnesiumbromide,bromoethylmagnesium,ethylmagnesium bromide solution, 1.0 m in thf,ethylmagnesium bromide solution, 3.0 m in diethyl ether,etmgbr,ethylmagnesiumbromid,ethylmagnesium bromid
101914
magnesium;ethane;bromide
C[CH2-].[Mg+2].[Br-]
925-90-6,96-47-9
C2H5BrMg
133.271
MFCD00000043
FRIJBUGBVQZNTB-UHFFFAOYSA-M
ethylmagnesium bromide,ethyl magnesium bromide,magnesium, bromoethyl,ethylmagnesiumbromide,bromoethylmagnesium,ethylmagnesium bromide solution, 1.0 m in thf,ethylmagnesium bromide solution, 3.0 m in diethyl ether,etmgbr,ethylmagnesiumbromid,ethylmagnesium bromid
101914
magnesium;ethane;bromide
C[CH2-].[Mg+2].[Br-]
Linear Formula CH3CH2MgBr
Concentration 39 to 41%
Fieser 02,205
Density 1.1400g/mL
Formula Weight 133.27
Physical Form Liquid
Packaging AcroSeal™ Glass bottle
Solubility Solubility in water: reacts.
Specific Gravity 1.14
Storage Information product may develop some precipitate or turbidity
Color Brown
Quantity 100mL
Flash Point −11°C
Chemical Name or Material Ethylmagnesium bromide, 3.2M solution in 2-Methyltetrahydrofuran
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Highly flammable liquid and vapor.
May cause respiratory irritation.
Reacts violently with water.
May form explosive peroxides.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger