Alfa Aesar™ Ethylmercury chloride

Manufacturer: Alfa Aesar™ 3711414

Quantity 25g

 View more versions of this product

Catalog No. AA3711414

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 107-27-7
    Linear Formula CH3CH2HgCl
    Formula Weight 265.13
    MDL Number MFCD00013593
    UN Number UN2025
    Merck Index 14,3826
    Synonym Chloroethylmercury; Ethyl mercuric chloride
    Chemical Name or Material Ethylmercury chloride

    Specifications

    Formula Weight 265.13
    Health Hazard 3 GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 193°C
    Physical Form Crystalline
    Solubility Information Insoluble in water; Slightly soluble in ether; Soluble in hot alcohol; Sublimes readily
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H373
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Color White
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, SOLID, N.O.S.
    RTECS Number OV9800000
    MDL Number MFCD00013593
    CAS 107-27-7
    Density 3.482g/mL
    UN Number UN2025
    Merck Index 14,3826
    Synonym Chloroethylmercury; Ethyl mercuric chloride
    Quantity 25g
    Linear Formula CH3CH2HgCl
    Health Hazard 1 Danger
    Chemical Name or Material Ethylmercury chloride

    GHS H Statement
    H300-H310-H330-H373
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    May cause damage to organs through prolonged or repeated exposure.

    GHS P Statement
    P260-P301+P310-P304+P340-P320-P330-P361-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit