Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ethylparaben, NF, 98-102%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
120-47-8
C9H10O3
166.18
NUVBSKCKDOMJSU-UHFFFAOYSA-N
ethyl 4-hydroxybenzoate
CCOC(=O)C1=CC=C(O)C=C1
120-47-8
C9H10O3
166.18
NUVBSKCKDOMJSU-UHFFFAOYSA-N
ethyl 4-hydroxybenzoate
CCOC(=O)C1=CC=C(O)C=C1

ET117, 120-47-8, C9H10O3

Spectrum™ Ethylparaben, NF is used as an antimicrobial preservative. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Packaging Poly Bottle
Ignition Residue 0.001
Quantity 500 g
CAS Max % 1
Percent Purity 98 to 102%