Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Ethylthio)trimethylsilane, 90%, ACROS Organics™

Click to view available options
Quantity:
1g
10g
Packaging:
Glass bottle
5573-62-6
C5H14SSi
134.3
MFCD00042890
HXAFQWICVBBXMZ-UHFFFAOYSA-N
ethylthio trimethylsilane,ethylthiotrimethylsilane,ethylthio trimethylsilane, technical grade,acmc-20aplk,trimethyl ethylthio silane,ethyl thio trimethylsilane,ethyl trimethylsilyl sulfide,trimethylsilyl ethyl sulfide,ethylsulfanyl trimethylsilane,ethylsulfanyl trimethyl silane
2733426
ethylsulfanyl(trimethyl)silane
CCS[Si](C)(C)C
5573-62-6
C5H14SSi
134.3
MFCD00042890
HXAFQWICVBBXMZ-UHFFFAOYSA-N
ethylthio trimethylsilane,ethylthiotrimethylsilane,ethylthio trimethylsilane, technical grade,acmc-20aplk,trimethyl ethylthio silane,ethyl thio trimethylsilane,ethyl trimethylsilyl sulfide,trimethylsilyl ethyl sulfide,ethylsulfanyl trimethylsilane,ethylsulfanyl trimethyl silane
2733426
ethylsulfanyl(trimethyl)silane
CCS[Si](C)(C)C
Quantity 1g
Density 0.8300g/mL
Refractive Index 1.4470 to 1.451
Flash Point 16°C
Boiling Point 69°C to 70°C (100.0mmHg)
Specific Gravity 0.83
Color Colorless
Infrared Spectrum Authentic
Packaging Glass bottle
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 88% min. (GC)
Linear Formula C2H5SSi(CH3)3
Formula Weight 134.3
Physical Form Liquid
Percent Purity 90%
Grade Technical
Chemical Name or Material (Ethylthio)trimethylsilane, 90%
Show More Show Less

GHS H Statement:
Highly flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

TSCA : TSCA