Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyltrimethoxysilane, 97%

Click to view available options
Quantity:
100g
5314-55-6
C5H14O3Si
150.249
MFCD00051541
SBRXLTRZCJVAPH-UHFFFAOYSA-N
silane, ethyltrimethoxy,ethyl trimethoxy silane,trimethoxyethylsilane,silane,ethyltrimethoxy,acmc-209l4b,ethyltrimethoxysilane,dow corning™ product 1-6321
79204
ethyl(trimethoxy)silane
CC[Si](OC)(OC)OC
5314-55-6
C5H14O3Si
150.249
MFCD00051541
SBRXLTRZCJVAPH-UHFFFAOYSA-N
silane, ethyltrimethoxy,ethyl trimethoxy silane,trimethoxyethylsilane,silane,ethyltrimethoxy,acmc-209l4b,ethyltrimethoxysilane,dow corning™ product 1-6321
79204
ethyl(trimethoxy)silane
CC[Si](OC)(OC)OC
UN Number UN1993
Formula Weight 150.25
Percent Purity 97%
Boiling Point 123°C to 124°C
Quantity 100g
Flash Point 27°C (81°F)
Chemical Name or Material Ethyltrimethoxysilane

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA