Alfa Aesar™ Ethyltriphenylphosphonium acetate, 70% in methanol

Manufacturer: Alfa Aesar™ 1742636

Quantity 500g

 View more versions of this product

Catalog No. AA1742636

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 35835-94-0
    Linear Formula CH3CH2P(C6H5)3O2CCH3
    Formula Weight 350.4
    MDL Number MFCD00061389
    UN Number UN1993
    Chemical Name or Material Ethyltriphenylphosphonium acetate

    Specifications

    Formula Weight 350.4
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Linear Formula CH3CH2P(C6H5)3O2CCH3
    MDL Number MFCD00061389
    Health Hazard 2 GHS H Statement
    H224-H300-H330-H315-H319-H335
    Extremely flammable liquid and vapor.
    Fatal if swallowed.
    Fatal if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Color Yellow to dark brown
    Quantity 500g
    TSCA TSCA
    Physical Form Liquid
    UN Number UN1993
    Health Hazard 1 Danger
    Density 1.05g/mL
    Concentration 70% in methanol
    Flash Point −17° (0°F)
    DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    CAS 35835-94-0
    Chemical Name or Material Ethyltriphenylphosphonium acetate

    GHS H Statement
    H224-H300-H330-H315-H319-H335
    Extremely flammable liquid and vapor.
    Fatal if swallowed.
    Fatal if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
    Rinse skin with water/shower.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Specific treatment is urgent (see label).
    Rinse mouth.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit