Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Ethyltriphenylphosphonium acetate, 70% in methanol

Manufacturer:  Alfa Aesar™ 1742622

Quantity 100g

 View more versions of this product

Catalog No. AA1742622

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

35835-94-0
350.4
UN1993
CH3CH2P(C6H5)3O2CCH3
MFCD00061389
Ethyltriphenylphosphonium acetate

Specifications

35835-94-0
CH3CH2P(C6H5)3O2CCH3
MFCD00061389
1.05g/mL
70% in methanol
−17° (0°F)
Liquid
350.4
UN1993
Yellow to dark brown
100g
Ethyltriphenylphosphonium acetate

Safety and Handling

SDS & Safety Documents

GHS H Statement
H224-H300-H330-H315-H319-H335
Extremely flammable liquid and vapor.
Fatal if swallowed.
Fatal if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit