Alfa Aesar™ Ethyltriphenylphosphonium bromide, 98+%

Manufacturer: Alfa Aesar™ B2309636

Quantity 500g

 View more versions of this product

Catalog No. AAB2309636

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 1530-32-1
    Molecular Formula C20H20BrP
    Linear Formula CH3CH2P(C6H5)3Br
    Formula Weight 371.26
    MDL Number MFCD00011838
    UN Number UN3077
    Beilstein 3599630
    Chemical Name or Material Ethyltriphenylphosphonium bromide

    Specifications

    Formula Weight 371.26
    Linear Formula CH3CH2P(C6H5)3Br
    MDL Number MFCD00011838
    Melting Point 206° to 210°C
    Flash Point >200°C (392°F)
    TSCA TSCA
    Molecular Formula C20H20BrP
    CAS 1530-32-1
    UN Number UN3077
    Beilstein 3599630
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Odor Odorless
    Quantity 500g
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    Chemical Name or Material Ethyltriphenylphosphonium bromide
    Assay ≥98%

    Description

    Applications
    Ethyltriphenylphosphonium bromide acts as a reactant in the synthesis of D-amino acids from L-cysteine-derived thiazolidines, Leiodolide A through aldol reactions and Horner-Wadsworth-Emmons olefination. It is also used in the preparation of cycloalkanoindolines through diastereoselective intramolecular inimo-ene reactions. Further, it is used as a reagent in solid-state metathesis polycondensation to prepare alkyl-dipropenylthiophene monomers and Mizoroki-Heck cyclization and cascading Tsuji-Trost cyclization reactions.

    Solubility
    Soluble in methanol. Slightly soluble in water, acetone and isopropanol.

    Notes
    Hygroscopic. Incompatible with oxidizing agents.

    GHS H Statement
    H302-H312-H315-H319-H335
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit