Alfa Aesar™ Ethyltriphenylphosphonium chloride, 98%

Manufacturer: Alfa Aesar™ B2301522

Quantity 100g

 View more versions of this product

Catalog No. AAB2301522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 896-33-3
    Molecular Formula CH3CH2P(C6H5)3Cl
    Formula Weight 326.81
    MDL Number MFCD00044699
    UN Number UN3077
    Beilstein 4170030
    Chemical Name or Material Ethyltriphenylphosphonium chloride

    Specifications

    Formula Weight 326.81
    TSCA TSCA
    Molecular Formula CH3CH2P(C6H5)3Cl
    MDL Number MFCD00044699
    UN Number UN3077
    Beilstein 4170030
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point ≈235°C (dec.)
    Quantity 100g
    DOT Information Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
    CAS 896-33-3
    Chemical Name or Material Ethyltriphenylphosphonium chloride
    Assay 98%

    GHS H Statement
    H301-H311-H315-H319-H335
    Toxic if swallowed.
    Toxic in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P301+P310-P305+P351+P338-P361-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit