Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Etidronic Acid, Approx. 60%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
Packaging:
Amber Glass Bottle
2809-21-4
C2H8O7P2
206.03
DBVJJBKOTRCVKF-UHFFFAOYSA-N
(1-hydroxy-1-phosphonoethyl)phosphonic acid
CC(O)(P(O)(O)=O)P(O)(O)=O
2809-21-4
C2H8O7P2
206.03
DBVJJBKOTRCVKF-UHFFFAOYSA-N
(1-hydroxy-1-phosphonoethyl)phosphonic acid
CC(O)(P(O)(O)=O)P(O)(O)=O

E3490, 2809-21-4, C2H8O7P2

Spectrum™ Etidronic Acid , also known as 1-hydroxyethane 1,1-diphosphonic acid, is used as a bisphosphonate to treat osteoporosis. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Amber Glass Bottle
Quantity 500 g
CAS Max % 1
Percent Purity Approx. 60%