Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Etodolac, Tocris Bioscience™

Catalog Number p-4816722
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
41340-25-4
C17H21NO3
287.36
MFCD00133313
NNYBQONXHNTVIJ-UHFFFAOYNA-N
etodolac, etodolic acid, lodine, ultradol, lodine xl, etodolaco, etodolacum, ramodar, etodolacum inn-latin, etodolaco inn-spanish
3308
CHEBI:4909
2-{1,8-diethyl-1H,3H,4H,9H-pyrano[3,4-b]indol-1-yl}acetic acid
CCC1=C2NC3=C(CCOC3(CC)CC(O)=O)C2=CC=C1
This item is not returnable. View return policy
41340-25-4
C17H21NO3
287.36
MFCD00133313
NNYBQONXHNTVIJ-UHFFFAOYNA-N
etodolac, etodolic acid, lodine, ultradol, lodine xl, etodolaco, etodolacum, ramodar, etodolacum inn-latin, etodolaco inn-spanish
3308
CHEBI:4909
2-{1,8-diethyl-1H,3H,4H,9H-pyrano[3,4-b]indol-1-yl}acetic acid
CCC1=C2NC3=C(CCOC3(CC)CC(O)=O)C2=CC=C1

Selective cyclooxygenase-2 (COX-2) inhibitor; NSAID

Non-steroidal anti-inflammatory drug (NSAID) that selectively inhibits cyclooxygenase-2 (COX-2) (IC50 values are 53 and >100 μM for COX-2 and COX-1 respectively). Displays anti-inflammatory effects in both adjuvant arthritic and normal rats.
Quantity 10 mg
Formula Weight 287.35
Percent Purity >98%
Chemical Name or Material Etodolac
Recommended Storage Store at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit