Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Etoposide, USP, 95-105%, Spectrum™

Click to view available options
Quantity:
25 g
Packaging:
Amber Glass Bottle

E2070, 33419-42-0, C29H32O13

Spectrum™ Etoposide, USP is an anticancer agent used alone or in combination with other medications to treat certain forms of lung cancer. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Ignition Residue 0.001
Water 0.06
Quantity 25 g
CAS 33419-42-0
CAS Max % 1
Molecular Formula C29H32O13
InChI Key VJJPUSNTGOMMGY-MRVIYFEKSA-N
SMILES COC1=CC(=CC(OC)=C1O)[C@H]1[C@@H]2[C@H](COC2=O)[C@H](O[C@@H]2O[C@@H]3CO[C@@H](C)O[C@H]3[C@H](O)[C@H]2O)C2=CC3=C(OCO3)C=C12
IUPAC Name (10R,11R,15R,16S)-16-{[(2R,4aR,6R,7R,8R,8aS)-7,8-dihydroxy-2-methyl-hexahydro-2H-pyrano[3,2-d][1,3]dioxin-6-yl]oxy}-10-(4-hydroxy-3,5-dimethoxyphenyl)-4,6,13-trioxatetracyclo[7.7.0.0³,⁷.0¹¹,¹⁵]hexadeca-1,3(7),8-trien-12-one
Molecular Weight (g/mol) 588.56
Percent Purity 95 to 105%
Show More Show Less