Learn More
ETS2 Mouse anti-Human, Clone: OTI1H4, liquid, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner TA505372
Catalog No. 50-167-5858
Add to cart
Description
Description
ETS transcriptions factors, such as ETS2, regulate numerous genes and are involved in stem cell development, cell senescence and death, and tumorigenesis. The conserved ETS domain within these proteins is a winged helix-turn-helix DNA-binding domain that recognizes the core consensus DNA sequence GGAA/T of target genes.
Specifications
Specifications
ETS2
Monoclonal
Unconjugated
ETS2
Liquid
ETS2
Human recombit protein fragment corresponding to amino acids 90-349 of human ETS2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI1H4
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
P15036
ETS2IT1
Mouse
IgG2b
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2114