Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ETS2 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen ETS2
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094924
View Documents
Novus Biologicals
NBP304070100UL
100 μg Each for $320.71
Add to cart
 
NB094925
View Documents
Novus Biologicals
NBP30407020UL
20 μg Each for $138.37
Add to cart
 
Description

Description

ETS2 Polyclonal antibody specifically detects ETS2 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

ETS2
Unconjugated
Rabbit
Stem Cell Markers
PBS with 50% glycerol, pH7.3.
2114
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Purified
RUO
Human, Mouse
ETS2IT1, human erythroblastosis virus oncogene homolog 210protein C-ets-2, oncogene ETS-2, v-ets avian erythroblastosis virus E2 oncogene homolog 2, v-ets avian erythroblastosis virus E26 oncogene homolog 2, v-ets erythroblastosis virus E26 oncogene homolog 2 (avian)
A synthetic peptide of human ETS2.
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit