Learn More
ETV3 Mouse anti-Human, Clone: 1C11, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524392
Catalog No. PIMA524392
Add to cart
Description
Description
Peptide Sequence: MKAGCSIVEK PEGGGGYQFP DWAYKTESSP GSRQIQLWHF ILELLQKEEF RHVIAWQQGE YGEFVIKDPD EVARLWGRRK CKPQMNYDKL SRALRYYYNK RILHKTKGKR FTYKFNFNKL VMPNYPFINI RSSGKIQTLL VGN
Transcriptional repressor that contribute to growth arrest during terminal macrophage differentiation by repressing target genes involved in Ras-dependent proliferation. Represses MMP1 promoter activity.
Specifications
Specifications
ETV3
Monoclonal
Unconjugated
ETV3
Liquid
ETV3
ETV3 (AAH22868, 1 a.a. ∽ 143 a.a) full-length recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Protein A
RUO
Antibody
Monoclonal
Human
ELISA
1C11
PBS with no preservative; pH 7.4
P41162
ETV3; METS; PE1
Mouse
IgG2b, kappa
100 μg
-20° C
Primary
2117