Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Etv3l Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570371
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%.
Transcriptional regulator.
Specifications
Etv3l | |
Polyclonal | |
Unconjugated | |
ETV3L | |
ETS translocation variant 3-like protein; ETS variant 3 like; ets variant 3-like; ets variant gene 3-like; ETS variant transcription factor 3 like; ETV3L; RGD1562620 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
499651 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
0 | |
ETV3L | |
synthetic peptide directed towards the middle region of rat Etv3l. | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction