Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ETV7 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS5965R

Description
The ETS (E26 transformation-specific) family of transcription factors, of which ETV7 is a member, is a large group of evolutionarily conserved transcriptional regulators that play an important role in a variety of cellular processes throughout development and differentiation, and are involved in oncogenesis as well (Potter et al., 2000 [PubMed 10828014]).Specifications
ETV7 | |
Polyclonal | |
Unconjugated | |
ETV7 | |
Ets transcription factor TEL-2b; ETS translocation variant 7; ETS variant 7; ets variant gene 7 (TEL2 oncogene); ETS-related protein Tel2; ETV7; TEL2; TEL-2; TEL2 oncogene; TELB; Tel-related Ets factor; Transcription factor ETV7; Transcription factor Tel-2; TREF | |
Rabbit | |
Protein A | |
RUO | |
51513 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9Y603 | |
ETV7 | |
KLH conjugated synthetic peptide derived from human ETV7. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction