Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eugenol 99%, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
2.5kg
Packaging:
Glass bottle
97-53-0
C10H12O2
164.2
MFCD00008654
RRAFCDWBNXTKKO-UHFFFAOYSA-N
eugenol,4-allylguaiacol,4-allyl-2-methoxyphenol,eugenic acid,allylguaiacol,caryophyllic acid,p-allylguaiacol,p-eugenol,2-methoxy-4-allylphenol,engenol
3314
CHEBI:4917
2-methoxy-4-prop-2-enylphenol
COC1=C(C=CC(=C1)CC=C)O
97-53-0
C10H12O2
164.2
MFCD00008654
RRAFCDWBNXTKKO-UHFFFAOYSA-N
eugenol,4-allylguaiacol,4-allyl-2-methoxyphenol,eugenic acid,allylguaiacol,caryophyllic acid,p-allylguaiacol,p-eugenol,2-methoxy-4-allylphenol,engenol
3314
CHEBI:4917
2-methoxy-4-prop-2-enylphenol
COC1=C(C=CC(=C1)CC=C)O
Melting Point -12°C to 10°C
Color Yellow
Refractive Index 1.5400 to 1.5420
Packaging Glass bottle
Boiling Point 254.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Specific Gravity 1.06
Quantity 500g
Density 1.0600g/mL
Assay Percent Range 99%
Linear Formula (H2C=CHCH2)C6H3(OCH3)OH
Beilstein 06,961
Merck Index 15,394
Solubility Information Solubility in water: 2.46g/L (25°C). Other solubilities: soluble in glacial acetic acid,soluble in alcohol,chloroform and ether
Formula Weight 164.2
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Eugenol, 99%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 202-589-1

RTECSNumber : SJ4375000

TSCA : TSCA