Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eukaryotic translation initiation factor 1 Mouse anti-Human, PE/Cy5.5, Clone: OTI2B9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271554PECY55

Catalog No. N27154PECY5


Add to cart

Description

Description

Eukaryotic translation initiation factor 1 Monoclonal antibody specifically detects Eukaryotic translation initiation factor 1 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

Eukaryotic translation initiation factor 1
OTI2B9
Flow Cytometry
Affinity Purified, Conjugated
EIF1
Full length recombinant protein of human Eukaryotic translation initiation factor 1 (NP_005792) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
PE-Cyanine5.5
PBS with 0.05% Sodium Azide
A121Protein translation factor SUI1 homolog, eIF1, EIF-1, EIF1A, eukaryotic translation initiation factor 1, ISO1, Sui1iso1, SUI1sui1iso1
Mouse
IgG2a
0.1 mL
Primary
10209.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit