Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eukaryotic translation initiation factor 1 Antibody (OTI2B9), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271554PECY7
Description
Eukaryotic translation initiation factor 1 Monoclonal specifically detects Eukaryotic translation initiation factor 1 in Human, Rat samples. It is validated for Flow Cytometry.Specifications
Eukaryotic translation initiation factor 1 | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
EIF1 | |
Full length recombinant protein of human Eukaryotic translation initiation factor 1 (NP_005792) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Rat | |
IgG2a |
Flow Cytometry | |
OTI2B9 | |
Flow Cytometry | |
A121Protein translation factor SUI1 homolog, eIF1, EIF-1, EIF1A, eukaryotic translation initiation factor 1, ISO1, Sui1iso1, SUI1sui1iso1 | |
Mouse | |
Affinity Purified | |
DNA replication Transcription Translation and Splicing | |
10209.0 | |
Store at 4°C in the dark. Do not freeze. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction