Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GSC Go Science Crazy Euro Style Polypropylene Beakers Products for Science Education

Click to view available options
Capacity (Metric):
100 mL
1000 mL
2000 mL
250 mL
50 mL
500 mL
Assorted
Intended for ages 13 and up.
Intended for ages 13 and up.

Incorporate raised outer graduations for ease of reading. GSC International Euro Style Polypropylene Beakers are tough, chemical resistant, and autoclavable. They are great additions to laboratory or classroom settings.

  • Molded from tough translucent polypropylene
  • Incorporate raised outer graduations for ease of reading
  • Tapered spout for easy pouring
  • Chemical resistant
  • Autoclavable
Recommended Applications Education
Autoclavable Yes
Color Translucent
Capacity (Metric) 100 mL
Material Polypropylene
Graduated Yes
Includes Tapered spout
Scale Raised graduations
Style Euro