Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Europium foil, 0.1mm (0.004 in.) thick, 99.9% (REO)

Manufacturer:  Alfa Aesar™ 13965FI

Quantity 50 x 50mm

 View more versions of this product

Catalog No. AA13965FI

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Chemical Identifiers

7440-53-1
MFCD00010992
UN3208
Europium foil, 0.1mm (0.004 in.) thick

Specifications

≈0.33g/25x25mm
50 x 50mm
Odorless
Foil
UN3208
99.9%, (REO)
Gray
7440-53-1
Mylar under Argon
MFCD00010992
Europium foil, 0.1mm (0.004 in.) thick

Safety and Handling

SDS & Safety Documents

GHS H Statement
H250-H261

GHS P Statement
P210-P222-P231+P232-P280-P233-P370+P378b-P335+P334-P422a-P402+P404-P501a

Danger

DOTInformation : Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.

TSCA : TSCA

storageNote1 :

  • Packaged under argon

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit