Alfa Aesar™ Europium foil, 0.25mm (0.01 in.) thick, 99.9% (REO)

Manufacturer: Alfa Aesar™ 10175FF

Quantity 25 x 25mm

 View more versions of this product

Catalog No. AA10175FF

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-53-1
    MDL Number MFCD00010992
    UN Number UN3208
    Chemical Name or Material Europium foil, 0.25mm (0.01 in.) thick

    Specifications

    Quantity 25 x 25mm
    Packaging Mylar under Argon
    Health Hazard 3 GHS P Statement
    P210-P222-P231+P232-P370+P378b-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not allow contact with air.
    Handle under inert gas.
    Protect from moisture.
    In case of fire: Use for
    Odor Odorless
    Physical Form Foil
    MDL Number MFCD00010992
    DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: METALLIC SUBSTANCE, WATER-REACTIVE, N.O.S.
    Weight ≈0.82g/25x25mm
    TSCA TSCA
    CAS 7440-53-1
    UN Number UN3208
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H250-H261
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gas.
    Storage Note 1
    • Packaged under argon
    Color Gray
    Chemical Name or Material Europium foil, 0.25mm (0.01 in.) thick
    Assay 99.9%, (REO)

    GHS H Statement
    H250-H261
    Catches fire spontaneously if exposed to air.
    In contact with water releases flammable gas.

    GHS P Statement
    P210-P222-P231+P232-P370+P378b-P422a-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    No smoking.
    Do not allow contact with air.
    Handle under inert gas.
    Protect from moisture.
    In case of fire: Use for

    • Packaged under argon

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit