Alfa Aesar™ Europium(III) acetate hydrate, REacton™, 99.999% (REO)

Manufacturer: Alfa Aesar™ 4405609

Quantity 10g

 View more versions of this product

Catalog No. AA4405609

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 62667-64-5
    Molecular Formula Eu(OOCCH3)3·xH2O
    Formula Weight 329.10 (Anhydrous)
    MDL Number MFCD00150115
    Chemical Name or Material Europium (III) acetate hydrate
    Grade Reacton™

    Specifications

    Formula Weight 329.10 (Anhydrous)
    Sensitivity Hygroscopic
    CAS 62667-64-5
    TSCA TSCA
    Physical Form Lumps/Powder
    Molecular Formula Eu(OOCCH3)3·xH2O
    Quantity 10g
    MDL Number MFCD00150115
    Chemical Name or Material Europium (III) acetate hydrate
    Assay 99.999%, (REO)
    Grade Reacton™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit