Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Europium(III) carbonate hydrate, 99.99% (REO), Alfa Aesar™, REacton™

Manufacturer:  Alfa Aesar 1321206

Quantity 5g

 View more versions of this product

Catalog No. AA1321206


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

86546-99-8
99.99% (REO)
483.95 (Anhydrous)
Europium(III) carbonate hydrate
Powder
Eu2(CO3)3·xH2O
MFCD00149702

Specifications

86546-99-8
99.99% (REO)
483.95 (Anhydrous)
MFCD00149702
White
Europium(III) carbonate hydrate
Powder
Eu2(CO3)3·xH2O
White powder
Odorless
5g

Safety and Handling

SDS & Safety Documents

EINECSNumber : 227-582-0

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit