Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Europium(III) chloride hexahydrate, 99.99%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
13759-92-7
Cl3Eu·6H2O
366.4
MFCD00149701
13759-92-7
Cl3Eu·6H2O
366.4
MFCD00149701
for ultra trace analysis
Assay Percent Range 39.0 to 42.5% (Eu) (Complexometry)
Linear Formula EuCl3·6H2O
Fieser 15,186
Merck Index 10, 3854
Solubility Information Solubility in water: soluble
Formula Weight 366.4
Physical Form Crystalline Powder, Crystals, and/or Chunks
Percent Purity 99.99%
Grade Trace Metal Basis
Density 4.8900g/mL
Packaging Glass bottle
Solubility Solubility in water: soluble.
Specific Gravity 4.89
Total Impurity 0.01% max.
Color White
Melting Point 850.0°C
Quantity 25g
Chemical Name or Material Europium(III) chloride hexahydrate
Show More Show Less