Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Europium(III) chloride, ultra dry, 99.99% (REO)

Click to view available options
Quantity:
1g
5g
25g
10025-76-0
Cl3Eu
258.314
MFCD00010994
NNMXSTWQJRPBJZ-UHFFFAOYSA-K
acmc-20akvq,europium 3+ trichloride,europium 3+ ion trichloride,europium iii chloride, ultra dry reo,europium chloride, anhydrous,europium iii chloride, anhydrous, ampuled under argon trace rare earth metals basis 1g
9859933
europium(3+);trichloride
[Cl-].[Cl-].[Cl-].[Eu+3]
10025-76-0
Cl3Eu
258.314
MFCD00010994
NNMXSTWQJRPBJZ-UHFFFAOYSA-K
acmc-20akvq,europium 3+ trichloride,europium 3+ ion trichloride,europium iii chloride, ultra dry reo,europium chloride, anhydrous,europium iii chloride, anhydrous, ampuled under argon trace rare earth metals basis 1g
9859933
europium(3+);trichloride
[Cl-].[Cl-].[Cl-].[Eu+3]
  • Ampouled under argon
Merck Index 14,3904
Solubility Information Soluble in water; Soluble in dilute hydrochloric acid,acetone and alcohol; Slightly soluble in tetrahydrofuran.
Formula Weight 258.32
Physical Form Powder
Percent Purity 99.99%
Packaging Ampule
Sensitivity Hygroscopic
Melting Point 850°C
Quantity 25g
Chemical Name or Material Europium(III) chloride, Ultra dry
Assay (REO)
Show More Show Less

EINECSNumber : 233-040-4

RTECSNumber : LE7525000

TSCA : Yes

storageNote1 :

  • Ampouled under argon

Recommended Storage : Ambient temperatures; Store under Nitrogen