Learn More
Europium(III) fluoride, anhydrous, REacton™, 99.9% (REO), Thermo Scientific Chemicals
Description
Europium(III) fluoride is used as a catalyst in the preparation of novel mixed-metal fluorides. It finds application in studies related to fluoride glasses. It is also used as a laboratory reagent. Further, it is used in optical glasses and ceramics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Boiling Point | >2280°C |
Melting Point | 1390°C |
Physical Form | Crystalline |
Quantity | 10 g |
Solubility Information | Slightly soluble in strong mineral acids. Insoluble in water. |
Formula Weight | 208.96 |
Percent Purity | 99.9% |
Grade | Reacton™ |
Sensitivity | Hygroscopic |
Assay | (REO) |
Show More |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 237-368-9 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.