Europium(III) nitrate hexahydrate, 99.9% (REO), Alfa Aesar™, Reacton™

Manufacturer: Alfa Aesar 1529009

Quantity 10g

 View more versions of this product

Catalog No. AA1529009

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10031-53-5
    Physical Form Crystalline
    Assay Percent Range 99.9% (REO)
    Molecular Formula Eu(NO3)3·6H2O
    Formula Weight 446.06; 265.90 (anhydrous)
    MDL Number MFCD00149700
    Chemical Name or Material Europium(III) nitrate hexahydrate

    Specifications

    Formula Weight 446.06; 265.90 (anhydrous)
    Quantity 10g
    Color Colorless
    EINECS Number 233-380-3
    Physical Form Crystalline
    Odor Odorless
    Assay Percent Range 99.9% (REO)
    Molecular Formula Eu(NO3)3·6H2O
    Appearance Colorless crystalline
    MDL Number MFCD00149700
    CAS 10031-53-5
    Health Hazard 1 H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
    Chemical Name or Material Europium(III) nitrate hexahydrate

    H272-H315-H319-H335: May intensify fire; oxidizer. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit