Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Europium(III) oxide, 99.9% (REO)

Click to view available options
Quantity:
2g
10g
1308-96-9
Eu2O3
351.925
MFCD00010997
AEBZCFFCDTZXHP-UHFFFAOYSA-N
europium oxide,europia,europium trioxide,dieuropium trioxide,europium sesquioxide,europium 3+ oxide,europium oxide euo1.5,europium 3+ ; oxygen 2-
159371
europium(3+);oxygen(2-)
[O-2].[O-2].[O-2].[Eu+3].[Eu+3]
1308-96-9
Eu2O3
351.925
MFCD00010997
AEBZCFFCDTZXHP-UHFFFAOYSA-N
europium oxide,europia,europium trioxide,dieuropium trioxide,europium sesquioxide,europium 3+ oxide,europium oxide euo1.5,europium 3+ ; oxygen 2-
159371
europium(3+);oxygen(2-)
[O-2].[O-2].[O-2].[Eu+3].[Eu+3]

For vacuum deposition

Merck Index 14,3904
Formula Weight 351.92
Physical Form Sintered Lump
Percent Purity 99.9%
Particle Size 3 to 12mm
Sensitivity Hygroscopic
Melting Point 2291°C
Quantity 10g
Chemical Name or Material Europium(III) oxide
Assay (REO)