Europium(III) oxide, 99.99% (REO), Alfa Aesar™, REacton™

Manufacturer: Alfa Aesar 1129906

Quantity 5g

 View more versions of this product

Catalog No. AA1129906

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1308-96-9
    Physical Form Powder or Chunks
    Assay Percent Range 99.99% (REO)
    Molecular Formula Eu2O3
    Formula Weight 351.92
    MDL Number MFCD00010997
    Chemical Name or Material Europium(III) oxide

    Specifications

    Physical Form Powder or Chunks
    Formula Weight 351.92
    Quantity 5g
    Color White
    Density 7.42g/cm3
    EINECS Number 215-165-6
    Assay Percent Range 99.99% (REO)
    Molecular Formula Eu2O3
    Appearance White powder or chunks
    MDL Number MFCD00010997
    CAS 1308-96-9
    Chemical Name or Material Europium(III) oxide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit