Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Europium(III) oxide, 99.99%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
1308-96-9
Eu2O3
351.91
MFCD00010997
AEBZCFFCDTZXHP-UHFFFAOYSA-N
europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2-
159371
europium(3+);oxygen(2-)
[O-2].[O-2].[O-2].[Eu+3].[Eu+3]
1308-96-9
Eu2O3
351.91
MFCD00010997
AEBZCFFCDTZXHP-UHFFFAOYSA-N
europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2-
159371
europium(3+);oxygen(2-)
[O-2].[O-2].[O-2].[Eu+3].[Eu+3]
for ultra trace analysis
Assay Percent Range 99.99%
Linear Formula Eu2O3
Merck Index 10, 3854
Solubility Information Solubility in water: insoluble
Formula Weight 351.91
Physical Form Powder
Percent Purity 99.99%
Grade Trace Metal Basis
Density 7.4200g/mL
Packaging Glass bottle
Solubility Solubility in water: insoluble.
Specific Gravity 7.42
Total Impurity 0.1% max.
Color White to Pink
Quantity 1g
Chemical Name or Material Europium(III) oxide
Show More Show Less

EINECSNumber : 215-165-6